missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hydroxysteroid (17-beta) dehydrogenase 11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | hydroxysteroid (17-beta) dehydrogenase 11 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
hydroxysteroid (17-beta) dehydrogenase 11 Polyclonal specifically detects hydroxysteroid (17-beta) dehydrogenase 11 in Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
| hydroxysteroid (17-beta) dehydrogenase 11 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| 17betaHSD11, 17betaHSDXI, 17-beta-hydroxysteroid dehydrogenase 11, 17-beta-hydroxysteroid dehydrogenase type XI, 17-beta-hydroxysteroid dehydrogenase XI, 17bHSD11, CTCL-associated antigen HD-CL-03, dehydrogenase/reductase (SDR family) member 8, DHRS8, EC 1.1.1, EC 1.1.1.62, hydroxysteroid (17-beta) dehydrogenase 11, member 2,17-beta-HSD 11, PAN1BRETSDR2,17-BETA-HSD11, retSDR2, RetSDR2,17-BETA-HSDXI, SDR16C2,17BHSD11, T-cell lymphoma-associated antigen HD-CL-03,17-beta-HSD XI | |
| HSD17B11 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Q8NBQ5 | |
| 51170 | |
| Synthetic peptides corresponding to HSD17B11 (hydroxysteroid (17-beta) dehydrogenase 11) The peptide sequence was selected from the N terminal of HSD17B11. Peptide sequence TGEIVLITGAGHGIGRLTAYEFAKLKSKLVLWDINKHGLEETAAKCKGLG. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title