missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hydroxysteroid (17-beta) dehydrogenase 11 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99361V
This item is not returnable.
View return policy
Description
hydroxysteroid (17-beta) dehydrogenase 11 Polyclonal antibody specifically detects hydroxysteroid (17-beta) dehydrogenase 11 in Human samples. It is validated for Western Blot
Specifications
| hydroxysteroid (17-beta) dehydrogenase 11 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| 17betaHSD11, 17betaHSDXI, 17-beta-hydroxysteroid dehydrogenase 11, 17-beta-hydroxysteroid dehydrogenase type XI, 17-beta-hydroxysteroid dehydrogenase XI, 17bHSD11, CTCL-associated antigen HD-CL-03, dehydrogenase/reductase (SDR family) member 8, DHRS8, EC 1.1.1, EC 1.1.1.62, hydroxysteroid (17-beta) dehydrogenase 11, member 2,17-beta-HSD 11, PAN1BRETSDR2,17-BETA-HSD11, retSDR2, RetSDR2,17-BETA-HSDXI, SDR16C2,17BHSD11, T-cell lymphoma-associated antigen HD-CL-03,17-beta-HSD XI | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human hydroxysteroid (17-beta) dehydrogenase 11. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 51170 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction