missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ hydroxysteroid (17-beta) dehydrogenase 11 293T Cell Transient Overexpression Lysate
Shop All Bio Techne ProductsSpecifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Lipid and Metabolism |
| Target | hydroxysteroid (17-beta) dehydrogenase 11 |
| Synonym | 17betaHSD11, 17betaHSDXI, 17-beta-hydroxysteroid dehydrogenase 11, 17-beta-hydroxysteroid dehydrogenase type XI, 17-beta-hydroxysteroid dehydrogenase XI, 17bHSD11, CTCL-associated antigen HD-CL-03, dehydrogenase/reductase (SDR family) member 8, DHRS8, EC 1.1.1, EC 1.1.1.62, hydroxysteroid (17-beta) dehydrogenase 11, member 2,17-beta-HSD 11, PAN1BRETSDR2,17-BETA-HSD11, retSDR2, RetSDR2,17-BETA-HSDXI, SDR16C2,17BHSD11, T-cell lymphoma-associated antigen HD-CL-03,17-beta-HSD XI |
| Gene Symbol | HSD17B11 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?