missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl methacrylate, mixture of isomers, 97+%, stab. with ca 0.02% 4-methoxyphenol

Product Code. 11419086
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
Unit Size:
1000mL
250mL
5000mL
This item is not returnable. View return policy

Product Code. 11419086

Brand: Thermo Scientific Alfa Aesar A17951.AU

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27813-02-1 | C7H12O3 | 144.17 g/mol

Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27813-02-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00004536
InChI Key ZMARGGQEAJXRFP-UHFFFAOYNA-N
Synonym 2-hydroxypropyl methacrylate, 2-hydroxypropylmethacrylate, hpma, beta-hydroxypropyl methacrylate, acryester hp, 2-hydroxypropyl 2-methylacrylate, poly 2-hydroxypropyl methacrylate, 2-hydroxypropyl 2-methyl-2-propenoate, 2-propenoic acid, 2-methyl-, 2-hydroxypropyl ester, 2-hpma
PubChem CID 13539
ChEBI CHEBI:53440
IUPAC Name 2-hydroxypropyl 2-methylprop-2-enoate
SMILES CC(CO)OC(=O)C(C)=C

Specifications

Melting Point 70°C (1mmHg)
Density 1.029
Boiling Point 205°C to 209°C
Flash Point 97°C (207°F)
Refractive Index 1.448
Quantity 1000 mL
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material Hydroxypropyl methacrylate, mixture of isomers, stabilized with ≈0.02% 4-methoxyphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxypropyl methacrylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2-Hydroxypropyl methacrylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.