missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized

Product Code. 10331351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10331351 1 kg Glass bottle 1kg
10095800 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10331351 Supplier Thermo Scientific Acros Supplier No. 412150010
  • ONLINE EXCLUSIVE PRICE
    £81.70
    £73.65 / 1kg
    Save £8.05
    9% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27813-02-1 | C7H12O3 | 144.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27813-02-1
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00004536
InChI Key ZMARGGQEAJXRFP-UHFFFAOYNA-N
Synonym 2-hydroxypropyl methacrylate, 2-hydroxypropylmethacrylate, hpma, beta-hydroxypropyl methacrylate, acryester hp, 2-hydroxypropyl 2-methylacrylate, poly 2-hydroxypropyl methacrylate, 2-hydroxypropyl 2-methyl-2-propenoate, 2-propenoic acid, 2-methyl-, 2-hydroxypropyl ester, 2-hpma
PubChem CID 13539
ChEBI CHEBI:53440
IUPAC Name 2-hydroxypropyl 2-methylprop-2-enoate
SMILES CC(CO)OC(=O)C(C)=C

Specifications

CAS Min % 97.0
CAS Max % 100.0
Density 1.0300g/mL
Boiling Point 240.0°C
Flash Point 101°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (Mixture of isomers) (GC)
Packaging Glass bottle
Linear Formula CH2=C(CH3)CO2CH2CH(OH)CH3
Refractive Index 1.4470 to 1.4490
Quantity 1 kg
Specific Gravity 1.03
Solubility Information Solubility in water: 130g/L (25°C). Other solubilities: miscible in almost all organic solvents
Stabilizer 150 to 300ppm MEHQ
Viscosity 9.9 mm2/s (20°C)
Formula Weight 144.17
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Hydroxypropyl methacrylate, 96.5%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxypropyl methacrylate, mixture of isomers, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.