missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl β-Cyclodextrin, MP Biomedicals™

Product Code. 11418831 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418831 1 g 1g
11428831 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418831 Supplier MP Biomedicals Supplier No. 0215354001
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Molecular biology and Cell biology
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94035-02-6
Molecular Formula C54H102O39
Molecular Weight (g/mol) 1375.371
InChI Key BZRWCHBEBFPRGF-UHFFFAOYSA-N
Synonym 2-hydroxypropy-.beta.-cyclodextrin, 2-hydroxypropylether-b-cyclodextrin, hydroxypropyl beta-cyclodextrin, tetrakis 1 propanol ; @cyclo-hepta-amylose, cyclo-hepta-amylose; tetrakis propanol
PubChem CID 44134771
SMILES CCCO.CCCO.CCCO.CCCO.C(C1C2C(C(C(O1)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)OC5C(OC(C(C5O)O)OC6C(OC(C(C6O)O)OC7C(OC(C(C7O)O)OC8C(OC(O2)C(C8O)O)CO)CO)CO)CO)CO)CO)O)O)O

Specifications

Color White
Quantity 1 g
Formula Weight 1454
Physical Form Solid
Chemical Name or Material Hydroxypropyl β-Cyclodextrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.