missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl Cellulose, TRC

Product Code. 30392662
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392662 1 g
30393069 10 g
30389268 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392662 Supplier Toronto Research Chemicals Supplier No. TRCH9526851G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9004-64-2
Synonym 2-Hydroxypropyl cellulose, AeroWhip, AeroWhip 630, AeroWhip 640, Aqualon Klucel L, Cellulose Hydroxypropyl Ether, Celny H, Celny HPC-L, Celny HPC-M, Celny HPC-SSL, Celny L, Celny M, Celny SL, E 463, G 4000HXL, H 0386, H 0473, H 0474, H 0475, HPC, HPC-E, HPC-E (cellulose derivative), HPC-EF-G, HPC-H, HPC-HF, HPC-L, HPC-L 100M, HPC-LE-G, HPC-LG, HPC-LR, HPC-M, HPC-MF, HPC-MG, HPC-S, HPC-S (cellulose derivative), HPC-SL, HPC-SSL, Hydroxypropyl Cellulose, Hydroxypropyl Cellulose Ether, Hydroxypropyl Ether of Cellulose, Hyprolose, JK 491, K 8913, K 9113, Klucel, Klucel E, Klucel EF, Klucel EXF, Klucel G, Klucel GF, Klucel H, Klucel HF, Klucel HXF, Klucel J, Klucel JF, Klucel L, Klucel LF, Klucel M, Klucel MCS, Klucel MF, Klucel Nutra D, L-HPC, L-HPC 11, L-HPC 32W, L-HPC LH 11, L-HPC-LH 21, L-HPC-LH 22, L-HPC-LH 31, L-HPC-LH 41, L-HPC-LH-B 1, L-HPCLH 10, LH 11, LH 20, LH 20 (cellulose derivative), LH 21, LH 22, LH 31, LH 32, LH 33, LH-B 1, LHPC 20, LHPC 21, Lacrisert, Nisso H, Nisso HPC, Nisso HPC-H, Nisso HPC-L, Nisso HPC-M, Nisso HPC-SL, Nisso HPC-SSL, Nisso M, O-(2-Hydroxypropyl)cellulose, O-(Hydroxypropyl)cellulose, Oxypropylated Cellulose, PM 50, PM 50 (polymer), R 1000, R 1000 (cellulose derivative)

Specifications

Quantity 1 g
Chemical Name or Material Hydroxypropylcellulose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.