missing translation for 'onlineSavingsMsg'
Få mere at vide

Hydroxypropyl Cellulose, TRC

Artikelnummer. 30389268
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
1 g
10 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity
30389268 100 g
30393069 10 g
30392662 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30389268 Leverandør Toronto Research Chemicals Leverandørnr. TRCH952685100G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 9004-64-2
Synonym 2-Hydroxypropyl cellulose, AeroWhip, AeroWhip 630, AeroWhip 640, Aqualon Klucel L, Cellulose Hydroxypropyl Ether, Celny H, Celny HPC-L, Celny HPC-M, Celny HPC-SSL, Celny L, Celny M, Celny SL, E 463, G 4000HXL, H 0386, H 0473, H 0474, H 0475, HPC, HPC-E, HPC-E (cellulose derivative), HPC-EF-G, HPC-H, HPC-HF, HPC-L, HPC-L 100M, HPC-LE-G, HPC-LG, HPC-LR, HPC-M, HPC-MF, HPC-MG, HPC-S, HPC-S (cellulose derivative), HPC-SL, HPC-SSL, Hydroxypropyl Cellulose, Hydroxypropyl Cellulose Ether, Hydroxypropyl Ether of Cellulose, Hyprolose, JK 491, K 8913, K 9113, Klucel, Klucel E, Klucel EF, Klucel EXF, Klucel G, Klucel GF, Klucel H, Klucel HF, Klucel HXF, Klucel J, Klucel JF, Klucel L, Klucel LF, Klucel M, Klucel MCS, Klucel MF, Klucel Nutra D, L-HPC, L-HPC 11, L-HPC 32W, L-HPC LH 11, L-HPC-LH 21, L-HPC-LH 22, L-HPC-LH 31, L-HPC-LH 41, L-HPC-LH-B 1, L-HPCLH 10, LH 11, LH 20, LH 20 (cellulose derivative), LH 21, LH 22, LH 31, LH 32, LH 33, LH-B 1, LHPC 20, LHPC 21, Lacrisert, Nisso H, Nisso HPC, Nisso HPC-H, Nisso HPC-L, Nisso HPC-M, Nisso HPC-SL, Nisso HPC-SSL, Nisso M, O-(2-Hydroxypropyl)cellulose, O-(Hydroxypropyl)cellulose, Oxypropylated Cellulose, PM 50, PM 50 (polymer), R 1000, R 1000 (cellulose derivative)

Tekniske data

Quantity 100 g
Chemical Name or Material Hydroxypropylcellulose
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.