missing translation for 'onlineSavingsMsg'
Learn More

Hydroxypropyl bis (N-Hydroxyethyl-p-phenylenediamine Hydrochloride) (>90%), TRC

Product Code. 30398011
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30398011 250 mg
30397878 25 mg
30392153 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398011 Supplier Toronto Research Chemicals Supplier No. TRCH939745250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128729-28-2
Molecular Formula C19H28N4O3 . 4HCl
Molecular Weight (g/mol) 506.3
Synonym 1,3-bis[(4-Aminophenyl)(2-hydroxyethyl)amino]-2-propanol Tetrahydrochloride
IUPAC Name 1,3-bis[4-amino-N-(2-hydroxyethyl)anilino]propan-2-ol;tetrahydrochloride
SMILES Cl.Cl.Cl.Cl.Nc1ccc(cc1)N(CCO)CC(O)CN(CCO)c2ccc(N)cc2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C19H28N4O3.4ClH/c20-15-1-5-17(6-2-15)22(9-11-24)13-19(26)14-23(10-12-25)18-7-3-16(21)4-8-18;;;;/h1-8,19,24-26H,9-14,20-21H2;4*1H
Formula Weight 504.1229
Chemical Name or Material Hydroxypropyl bis (N-Hydroxyethyl-p-phenylenediamine Hydrochloride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.