missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyprogesterone Caproate, TRC
Specifications
Specifications
| Chemical Name or Material | Hydroxyprogesterone Caproate |
| CAS | 630-56-8 |
| Molecular Formula | C27 H40 O4 |
| Quantity | 50 mg |
| Synonym | Hydroxyprogesterone Caproate, Pregn-4-ene-3,20-dione, 17-hydroxy-, hexanoate (7CI,8CI), Progesterone, 17-hydroxy-, hexanoate (6CI), Hexanoic acid, ester with 17-hydroxypregn-4-ene-3,20-dione (8CI), 17α-Caproyloxy-P4, 17α-Caproyloxypregn-4-ene-3,20-dione, 17α-Caproyloxyprogesterone, 17α-Hydroxypregn-4-ene-3,20-dione caproate, 17α-Hydroxypregn-4-ene-3,20-dione hexanoate, 17α-Hydroxyprogesterone caproate, 17α-Hydroxyprogesterone hexanoate, 17α-Hydroxyprogesterone n-caproate, 3,20-Dioxopregn-4-en-17α-yl caproate, Delalutin, Depo-proluton, HPC, Hormofort, Hydroxyprogesterone caproate, Hydroxyprogesterone hexanoate, Hyproval PA, Lewntogest, NSC 17592, Neolutin, Pharlon, Primolut Depot, Procyte Depo, Proge, Progesterone caproate, Proluton Depot, Syngynon, Teralutil |
| InChI Formula | InChI=1S/C27H40O4/c1-5-6-7-8-24(30)31-27(18(2)28)16-13-23-21-10-9-19-17-20(29)11-14-25(19,3)22(21)12-15-26(23,27)4/h17,21-23H,5-16H2,1-4H3/t21-,22+,23+,25+,26+,27+/m1/s1 |
| SMILES | CCCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] hexanoate |
| Molecular Weight (g/mol) | 428.6 |
| Formula Weight | 428.2927 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?