missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyprogesterone Caproate, TRC

Product Code. 30390990
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390990 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30390990 Supplier Toronto Research Chemicals Supplier No. TRCH952490100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxyprogesterone Caproate
CAS 630-56-8
Molecular Formula C27 H40 O4
Quantity 100 mg
Synonym Hydroxyprogesterone Caproate, Pregn-4-ene-3,20-dione, 17-hydroxy-, hexanoate (7CI,8CI), Progesterone, 17-hydroxy-, hexanoate (6CI), Hexanoic acid, ester with 17-hydroxypregn-4-ene-3,20-dione (8CI), 17α-Caproyloxy-P4, 17α-Caproyloxypregn-4-ene-3,20-dione, 17α-Caproyloxyprogesterone, 17α-Hydroxypregn-4-ene-3,20-dione caproate, 17α-Hydroxypregn-4-ene-3,20-dione hexanoate, 17α-Hydroxyprogesterone caproate, 17α-Hydroxyprogesterone hexanoate, 17α-Hydroxyprogesterone n-caproate, 3,20-Dioxopregn-4-en-17α-yl caproate, Delalutin, Depo-proluton, HPC, Hormofort, Hydroxyprogesterone caproate, Hydroxyprogesterone hexanoate, Hyproval PA, Lewntogest, NSC 17592, Neolutin, Pharlon, Primolut Depot, Procyte Depo, Proge, Progesterone caproate, Proluton Depot, Syngynon, Teralutil
InChI Formula InChI=1S/C27H40O4/c1-5-6-7-8-24(30)31-27(18(2)28)16-13-23-21-10-9-19-17-20(29)11-14-25(19,3)22(21)12-15-26(23,27)4/h17,21-23H,5-16H2,1-4H3/t21-,22+,23+,25+,26+,27+/m1/s1
SMILES CCCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
IUPAC Name [(8R,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] hexanoate
Molecular Weight (g/mol) 428.6
Formula Weight 428.2927
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.