missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyprogesterone caproate, MedChemExpress

Product Code. p-200097433
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16461048

Brand: Medchem Express HYB0742

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hydroxyprogesterone caproate is a synthetic, steroidal progestin; an ester derivative of 17α-hydroxyprogesterone formed from caproic acid.

Hydroxyprogesterone caproate is a synthetic, steroidal progestin

Specifications

Chemical Name or Material Hydroxyprogesterone caproate
Physical Form Solid
CAS 630-56-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C27H40O4
Synonym 17α-Hydroxyprogesterone hexanoate 17α-Hydroxyprogesterone caproate
Solubility Information DMSO : 50 mg/mL (116.66 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.23 mM; Need ultrasonic)
SMILES CC([C@@]1(OC(CCCCC)=O)CC[C@@]2([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])CC[C@]12C)=O)=O
Molecular Weight (g/mol) 428.6
Formula Weight 428.6
Percent Purity 99.68%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.