missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxypivalic Acid Methyl Ester, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Hydroxypivalic Acid Methyl Ester |
| CAS | 14002-80-3 |
| Molecular Formula | C6 H12 O3 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C6H12O3/c1-6(2,4-7)5(8)9-3/h7H,4H2,1-3H3 |
| SMILES | COC(=O)C(C)(C)CO |
| IUPAC Name | methyl 3-hydroxy-2,2-dimethylpropanoate |
| Molecular Weight (g/mol) | 132.158 |
| Formula Weight | 132.079 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction