missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyphosphonoacetic Acid - 50% in water, TRC
Specifications
Specifications
| Chemical Name or Material | Hydroxyphosphonoacetic Acid |
| CAS | 23783-26-8 |
| Molecular Formula | C2 H5 O6 P |
| Quantity | 500 mg |
| Synonym | 2-Hydroxy-2-phosphonoacetic Acid, 2-Hydroxyphosphonoacetic Acid, 2-Hydroxyphosphonocarboxylic Acid, Belcor 575, HPPA (scale inhibitor), Hydroxyphosphonoacetic Acid, Phosphonoglycolic Acid, Phosphonohydroxyacetic Acid, α-Hydroxyphosphonoacetic Acid |
| InChI Formula | InChI=1S/C2H5O6P/c3-1(4)2(5)9(6,7)8/h2,5H,(H,3,4)(H2,6,7,8) |
| SMILES | OC(C(=O)O)P(=O)(O)O |
| IUPAC Name | 2-hydroxy-2-phosphonoacetic acid |
| Molecular Weight (g/mol) | 156.031 |
| Formula Weight | 155.982 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?