missing translation for 'onlineSavingsMsg'
Learn More

Hydroxymethylferrocene, 97%, Thermo Scientific™

Product Code. 11333658
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11333658 1 g 1g
11343658 5 g 5g
11323658 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11333658 Supplier Thermo Scientific Alfa Aesar Supplier No. 039257.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1273-86-5
Molecular Formula C11H12FeO
Molecular Weight (g/mol) 216.06
MDL Number MFCD00040878
InChI Key KLMBXXGIFRROFL-UHFFFAOYSA-N
Synonym Ferrocenemethanol
SMILES [Fe].c1cccc1.OCc1cccc1

Specifications

Melting Point 79°C to 81°C
Color Yellow
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information It is Soluble in water (partly miscible),and methanol.
Formula Weight 216.07
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Hydroxymethylferrocene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.