missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxymethoxy-N-desmethyl Toremifene (>80%), TRC
Specifications
Specifications
| Chemical Name or Material | Hydroxymethoxy-N-desmethyl Toremifene |
| Analyte or Component Names | Hydroxymethoxy-N-desmethyl Toremifene |
| Purity Grade Notes | HPLC |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H28 Cl N O3 |
| Synonym | Hydroxymethoxy-N-desmethyl Toremifene (>80%) |
| InChI Formula | InChI=1S/C26H28ClNO3/c1-28-17-18-31-24-13-7-21(8-14-24)26(20-3-9-22(29)10-4-20)25(15-16-27)19-5-11-23(30-2)12-6-19/h3-14,28-29H,15-18H2,1-2H3/b26-25- |
| Solution Type | Neat |
| SMILES | CNCCOc1ccc(cc1)\C(=C(\CCCl)/c2ccc(OC)cc2)\c3ccc(O)cc3 |
| IUPAC Name | 4-[(Z)-4-chloro-2-(4-methoxyphenyl)-1-[4-[2-(methylamino)ethoxy]phenyl]but-1-enyl]phenol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?