missing translation for 'onlineSavingsMsg'
Learn More

Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite (dry wt.)

Product Code. 11422368
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422368 500 g 500g
11432368 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422368 Supplier Thermo Scientific Alfa Aesar Supplier No. A15731.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 149-44-0 | CH3NaO3S | 118.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-44-0
Molecular Formula CH3NaO3S
Molecular Weight (g/mol) 118.08
MDL Number MFCD00040426
InChI Key XWGJFPHUCFXLBL-UHFFFAOYSA-M
Synonym sodium hydroxymethanesulfinate, rongalite, rongalit, aldanil, discolite, formapon, formopan, hydrolit, leptacid, leptacit
PubChem CID 23689980
SMILES [Na+].OCS([O-])=O

Specifications

Melting Point 64°C to 68°C
Quantity 500 g
Beilstein 3630814
Merck Index 14,8620
IUPAC Name sodium;hydroxymethanesulfinate
Formula Weight 118.09 (Anhydrous)
Percent Purity 85%
Grade Technical
Assay cont. up to 5% sodium sulfite (dry wt.)
Chemical Name or Material Hydroxymethanesulfinic acid sodium salt hydrate
compliance-icons
Product Identifier
  • Hydroxymethanesulfinic acid sodium salt hydrate, tech 85%, cont. up to 5% sodium sulfite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydroxymethanesulphinic acid sodium salthydrate, Sodium sulphite

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.