missing translation for 'onlineSavingsMsg'
Learn More

Hydroxymalonic Acid, TRC

Product Code. 30393276
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393276 50 mg 50mg
30396716 10 mg -
30395985 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393276 Supplier Toronto Research Chemicals Supplier No. TRCH53155050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-69-3
Molecular Formula C3H4O5
Molecular Weight (g/mol) 120.06
SMILES C(C(=O)O)(C(=O)O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C3H4O5/c4-1(2(5)6)3(7)8/h1,4H,(H,5,6)(H,7,8)
Formula Weight 120.0059
Chemical Name or Material Hydroxymalonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.