missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylapatite, fast flow

Product Code. 15432919
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432919 10 g 10g
15442919 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15432919 Supplier Thermo Scientific Alfa Aesar Supplier No. J60076.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids | CAS: 1306-06-5 | Ca5HO13P3 | 502.31 g/mol

Used for fractionation and purification of proteins, enzymes, plasmids and nucleic acids

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-06-5
Molecular Formula Ca5HO13P3
Molecular Weight (g/mol) 502.31
MDL Number MFCD00010904
InChI Key XYJRXVWERLGGKC-UHFFFAOYSA-D
Synonym Calcium phosphate hydroxide; Durapatite
IUPAC Name pentacalcium hydroxide triphosphate
SMILES [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

Color White
Linear Formula Ca5(OH)(PO4)3
Quantity 10 g
Merck Index 14,3467
Formula Weight 502.32
Physical Form Powder
Chemical Name or Material Hydroxylapatite, fast flow

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.