missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylapatite, for analysis

Product Code. 10446583 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10446583 100 g Glass bottle 100g
10199372 25 g Glass bottle 25g
10110273 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10446583 Supplier Thermo Scientific Acros Supplier No. 371261000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 1306-06-5 | Ca5HO13P3 | 502.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-06-5
Molecular Formula Ca5HO13P3
Molecular Weight (g/mol) 502.31
MDL Number MFCD00010904
InChI Key XYJRXVWERLGGKC-UHFFFAOYSA-D
Synonym Calcium phosphate hydroxide
IUPAC Name pentacalcium hydroxide triphosphate
SMILES [OH-].[Ca++].[Ca++].[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

Specifications

Melting Point 1700.0°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula 3Ca3(PO4)2·Ca(OH)2
Merck Index 15, 3514
Solubility Information Solubility in water: negligible
Formula Weight 502.31
Percent Purity 35 to 40% (Ca)
Grade Analytical
Loss on Ignition 8% max. (800°C)
Packaging Glass bottle
Chemical Name or Material Hydroxylapatite
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.