missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Sulfate, TRC

Product Code. 30391346
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391346 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391346 Supplier Toronto Research Chemicals Supplier No. TRCH9438301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxylamine Sulfate
CAS 10039-54-0
Molecular Formula H8N2O6S
Quantity 1 g
Synonym Bis(hydroxylamine) Sulfate, Bis(hydroxylammonium) Sulfate, Di(hydroxylamine) Sulfate, Envision MLB 2030, Hydroxyl Ammonium Sulfate ((HONH3)2SO4), Hydroxylamine Neutral Sulfate, Hydroxylammonium Sulfate, Lanasane LAB, Oxammonium Sulfate
InChI Formula InChI=1S/2H4NO.H2O4S/c2*1-2;1-5(2,3)4/h2*2H,1H3;(H2,1,2,3,4)/q2*+1;/p-2
SMILES NO.NO.OS(=O)(=O)O
IUPAC Name hydroxylamine;sulfuric acid
Molecular Weight (g/mol) 164.14
Formula Weight 164.0103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.