missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Solution (50 wt. % in H2O), TRC

Product Code. 30009651
Change view
Click to view available options
Quantity:
1 mL
2.5 mL
Unit Size:
1mL
2.5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009651 2.5 mL 2.5mL
30009695 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009651 Supplier Toronto Research Chemicals Supplier No. TRCH9437082.5ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7803-49-8
Molecular Formula H3 N O
Molecular Weight (g/mol) 33.03
Synonym Hydroxylamine
IUPAC Name hydroxylamine
SMILES NO

Specifications

Analyte or Component Names Hydroxylamine
Shipping Condition Room Temperature
Concentration 50%
InChI Formula InChI=1S/H3NO/c1-2/h2H,1H2
Solution Type Single Solution
Solvent or Matrix Water
Industry Type Forensics
Quantity 2.5 mL
Formula Weight 33.0215
Concentration or Composition (by Analyte or Components) Hydroxylamine 50%
Chemical Name or Material Hydroxylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.