missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine-O-sulfonic Acid, TRC

Product Code. 30392587
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392587 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392587 Supplier Toronto Research Chemicals Supplier No. TRCH9437655G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxylamine-O-sulfonic Acid
CAS 2950-43-8
Molecular Formula H3NO4S
Quantity 5 g
Synonym Amidoperoxymonosulfuric Acid (HOS(O)2ONH2), Amidosulfonic Peracid, Aminomonopersulfuric Acid, Ammonia-N-oxide-O-sulfonic Acid Betaine, HAOS, O-Sulfohydroxylamine, Hydroxylaminesulfonic Acid, Permonosulfamic Acid, Sulfoperamidic Acid, Sulfuric Acid Monoazanyl Ester
InChI Formula InChI=1S/H3NO4S/c1-5-6(2,3)4/h1H2,(H,2,3,4)
SMILES NOS(=O)(=O)O
IUPAC Name amino hydrogen sulfate
Molecular Weight (g/mol) 113.09
Formula Weight 112.9783
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.