missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxylamine-O-sulfonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Hydroxylamine-O-sulfonic Acid |
| CAS | 2950-43-8 |
| Molecular Formula | H3NO4S |
| Quantity | 25 g |
| Synonym | Amidoperoxymonosulfuric Acid (HOS(O)2ONH2), Amidosulfonic Peracid, Aminomonopersulfuric Acid, Ammonia-N-oxide-O-sulfonic Acid Betaine, HAOS, O-Sulfohydroxylamine, Hydroxylaminesulfonic Acid, Permonosulfamic Acid, Sulfoperamidic Acid, Sulfuric Acid Monoazanyl Ester |
| InChI Formula | InChI=1S/H3NO4S/c1-5-6(2,3)4/h1H2,(H,2,3,4) |
| SMILES | NOS(=O)(=O)O |
| IUPAC Name | amino hydrogen sulfate |
| Molecular Weight (g/mol) | 113.09 |
| Formula Weight | 112.9783 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?