missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, ReagentPlus™, Honeywell

Product Code. 15671520 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 kg
Unit Size:
100g
1kg
3kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15671520 100 g 100g
15601530 500 g 500g
15681520 1 kg 1kg
15691520 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15671520 Supplier Honeywell Research Chemicals Supplier No. 159417100G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5470-11-1
Molecular Formula ClH4NO
Molecular Weight (g/mol) 69.49
MDL Number MFCD00051089
InChI Key WTDHULULXKLSOZ-UHFFFAOYSA-N
Synonym hydroxylamine hydrochloride, hydroxylamine hcl, hydroxylammonium chloride, oxammonium hydrochloride, hydroxylamine, hydrochloride, hydroxyamine hydrochloride, hydroxylamine chloride 1:1, hydroxylamine, hydrochloride 1:1, hydroxyamine hcl, oxammonium hci
PubChem CID 443297
ChEBI CHEBI:5807
IUPAC Name hydroxylamine hydrochloride
SMILES Cl.NO

Specifications

Quantity 100 g
Linear Formula NH2OH · HCl
UN Number 2923
Beilstein 3539763
Formula Weight 69.49g/mol
Percent Purity 99%
Grade ReagentPlus
Density 1.67g/mL at 25°C
Chemical Name or Material Hydroxylamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.