missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride-15N, TRC

Product Code. 30394282
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394282 250 mg -
30396092 100 mg -
30403679 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394282 Supplier Toronto Research Chemicals Supplier No. TRCH943707250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40711-48-6
Molecular Formula Cl H . H3 15N O
Molecular Weight (g/mol) 70.48
Synonym Hydroxylamine-15N, hydrochloride (9CI), 15N-Hydroxylamine hydrochloride, [15N]Hydroxylamine hydrochloride, Hydroxylamine-15N hydrochloride
SMILES Cl.[15NH2]O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/ClH.H3NO/c;1-2/h1H;2H,1H2/i;1+1
Formula Weight 69.9952
Chemical Name or Material Hydroxylamine Hydrochloride-15N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.