missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyhexamide, MedChemExpress

Product Code. 16448668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448668 10 mg 10mg
16441098 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448668 Supplier Medchem Express Supplier No. HYB1103
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Hydroxyhexamide is a pharmacologically active metabolite of Acetohexamide, used as a hypoglycemic agents.

Hydroxyhexamide is a pharmacologically active metabolite of Acetohexamide, used as a hypoglycemic agents.

Specifications

Chemical Name or Material Hydroxyhexamide
CAS 3168-01-2
Color White
Purity Grade Notes Research
Molecular Formula C15H22N2O4S
Quantity 10 mg
Synonym (±)-Hydroxyhexamid
Solubility Information DMSO : ≥ 300 mg/mL (919.09 mM)
SMILES O=S(C1=CC=C(C(O)C)C=C1)(NC(NC2CCCCC2)=O)=O
Molecular Weight (g/mol) 326.41
Formula Weight 326.41
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.