missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyfasudil, MedChemExpress

Product Code. 16475908
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475908 10 mg 10mg
16462057 50 mg 50mg
16448547 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16475908 Supplier Medchem Express Supplier No. HY13911

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hydroxyfasudil is a ROCK inhibitor, with IC50s of 0.73 and 0.72 μM for ROCK1 and ROCK2, respectively.

Hydroxyfasudil is a ROCK inhibitor, with IC50s of 0.73 and 0.72 μM for ROCK1 and ROCK2, respectively.

Specifications

Chemical Name or Material Hydroxyfasudil
CAS 105628-72-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C14H17N3O3S
Quantity 10 mg
Synonym HA-1100
Solubility Information DMSO : ≥ 31 mg/mL (100.86 mM)
SMILES O=C1NC=CC2=C1C=CC=C2S(=O)(N3CCNCCC3)=O
Molecular Weight (g/mol) 307.37
Formula Weight 307.37
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.