missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyethoxy Ethylene Terephthalate, TRC

Product Code. 30398081
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398081 10 mg 10mg
30402512 5 mg 5mg
30403057 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30398081 Supplier Toronto Research Chemicals Supplier No. TRCH95298110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxyethoxy Ethylene Terephthalate
Analyte or Component Names Hydroxyethoxy Ethylene Terephthalate
Shipping Condition Room Temperature
Molecular Formula C31 H30 O12
Synonym 2-((4-((2-Hydroxyethoxy)carbonyl)benzoyl)oxy)ethyl (2-((4-(methoxymethyl)benzoyl)oxy)ethyl) terephthalate
InChI Formula InChI=1S/C31H30O12/c1-38-20-21-2-4-22(5-3-21)28(34)40-16-17-41-30(36)25-10-12-26(13-11-25)31(37)43-19-18-42-29(35)24-8-6-23(7-9-24)27(33)39-15-14-32/h2-13,32H,14-20H2,1H3
Solution Type Neat
SMILES COCc1ccc(cc1)C(=O)OCCOC(=O)c2ccc(cc2)C(=O)OCCOC(=O)c3ccc(cc3)C(=O)OCCO
IUPAC Name 4-O-(2-hydroxyethyl) 1-O-[2-[4-[2-[4-(methoxymethyl)benzoyl]oxyethoxycarbonyl]benzoyl]oxyethyl] benzene-1,4-dicarboxylate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 594.563
Quantity 10 mg
Formula Weight 594.174
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.