missing translation for 'onlineSavingsMsg'
Learn More

Hydroxydiazinon, TRC

Product Code. 30391620
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391620 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391620 Supplier Toronto Research Chemicals Supplier No. TRCH21500010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxydiazinon
CAS 29820-16-4
Molecular Formula C12 H21 N2 O4 P S
Quantity 10 mg
Synonym Phosphorothioic acid, O,O-diethyl O-[2-(1-hydroxy-1-methylethyl)-6-methyl-4-pyrimidinyl] ester, 2-Pyrimidinemethanol, 4-hydroxy-α,α,6-trimethyl-, 4-O-ester with O,O-diethyl phosphorothioate (8CI), O,O-Diethyl O-[2-(1-hydroxy-1-methylethyl)-6-methylpyrimidin-4-yl] phosphorothioate, O,O-Diethyl O-[2-(2-hydroxyisopropyl)-6-methyl-4-pyrimidinyl] phosphorothioate, O,O-Diethyl O-[2-(2'-hydroxy-2'-propyl)-4-methyl-6-pyrimidinyl]phosphorothioate
InChI Formula InChI=1S/C12H21N2O4PS/c1-6-16-19(20,17-7-2)18-10-8-9(3)13-11(14-10)12(4,5)15/h8,15H,6-7H2,1-5H3
SMILES CCOP(=S)(OCC)Oc1cc(C)nc(n1)C(C)(C)O
IUPAC Name 2-(4-diethoxyphosphinothioyloxy-6-methylpyrimidin-2-yl)propan-2-ol
Molecular Weight (g/mol) 320.345
Formula Weight 320.096
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.