missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine Sulphate 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30461443
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461443 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461443 Supplier LoGiCal Supplier No. LGCAMP0764.0001
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxychloroquine sulfate
CAS 747-36-4
Analyte or Component Names Hydroxychloroquine sulfate
Shipping Condition Room Temperature
Molecular Formula C18 H26 Cl N3 O . H2 O4 S
Certifications/Compliance ISO 17034
Synonym Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]ethylamino]-, sulfate (1:1), Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]ethylamino]-, sulfate (1:1) (salt) (9CI), Ethanol, 2-[[4-[(7-chloro-4-quinolyl)amino]pentyl]ethylamino]-, sulfate (1:1) (salt) (8CI), 7-Chloro-4-[4-N-ethyl-N-beta-hydroxyethylamino-1-methylbutylamino]quinoline sulfate, 7-Chloro-4-[4-[ethyl(2-hydroxyethyl)amino]-1-methylbutylamino]-quinoline sulfate, Ercoquin, Hydroxychloroquine sulfate, NSC 4375, Plaquenil, Plaquenil sulfate, Quensyl
InChI Formula InChI=1S/C18H26ClN3O.H2O4S/c1-3-22(11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18;1-5(2,3)4/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21);(H2,1,2,3,4)
Solution Type Single Solution
SMILES CCN(CCO)CCCC(C)Nc1ccnc2cc(Cl)ccc12.OS(=O)(=O)O
IUPAC Name 2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 433.95
Quantity 1 mL
Formula Weight 433.1438 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.