missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine Sulfate, TRC

Product Code. 30009421
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009421 25 mg 25mg
30008077 10 mg 10mg
30009642 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009421 Supplier Toronto Research Chemicals Supplier No. TRCH91690025MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 747-36-4
Molecular Formula C18 H26 Cl N3 O . H2 O4 S
Molecular Weight (g/mol) 433.95
Synonym Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]ethylamino]-, sulfate (1:1), Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]ethylamino]-, sulfate (1:1) (salt) (9CI), Ethanol, 2-[[4-[(7-chloro-4-quinolyl)amino]pentyl]ethylamino]-, sulfate (1:1) (salt) (8CI), 7-Chloro-4-[4-N-ethyl-N-beta-hydroxyethylamino-1-methylbutylamino]quinoline sulfate, 7-Chloro-4-[4-[ethyl(2-hydroxyethyl)amino]-1-methylbutylamino]-quinoline sulfate, Ercoquin, Hydroxychloroquine sulfate, NSC 4375, Plaquenil, Plaquenil sulfate, Quensyl
IUPAC Name 2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid
SMILES CCN(CCO)CCCC(C)Nc1ccnc2cc(Cl)ccc12.OS(=O)(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H26ClN3O.H2O4S/c1-3-22(11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18;1-5(2,3)4/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21);(H2,1,2,3,4)
Formula Weight 433.1438
Percent Purity >95%
Chemical Name or Material Hydroxychloroquine sulfate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.