missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine Sulfate, TRC

Product Code. 30394855
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394855 250 mg 250mg
30404259 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394855 Supplier Toronto Research Chemicals Supplier No. TRCH916900250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxychloroquine sulfate
CAS 747-36-4
Purity Grade Notes HPLC
Molecular Formula C18 H26 Cl N3 O . H2 O4 S
Quantity 250 mg
Synonym Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]ethylamino]-, sulfate (1:1), Ethanol, 2-[[4-[(7-chloro-4-quinolinyl)amino]pentyl]ethylamino]-, sulfate (1:1) (salt) (9CI), Ethanol, 2-[[4-[(7-chloro-4-quinolyl)amino]pentyl]ethylamino]-, sulfate (1:1) (salt) (8CI), 7-Chloro-4-[4-N-ethyl-N-beta-hydroxyethylamino-1-methylbutylamino]quinoline sulfate, 7-Chloro-4-[4-[ethyl(2-hydroxyethyl)amino]-1-methylbutylamino]-quinoline sulfate, Ercoquin, Hydroxychloroquine sulfate, NSC 4375, Plaquenil, Plaquenil sulfate, Quensyl
InChI Formula InChI=1S/C18H26ClN3O.H2O4S/c1-3-22(11-12-23)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18;1-5(2,3)4/h6-9,13-14,23H,3-5,10-12H2,1-2H3,(H,20,21);(H2,1,2,3,4)
SMILES CCN(CCO)CCCC(C)Nc1ccnc2cc(Cl)ccc12.OS(=O)(=O)O
IUPAC Name 2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid
Molecular Weight (g/mol) 433.95
Formula Weight 433.1438
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.