missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine sulfate, 98%

Product Code. 10209543 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10209543 100 g Glass bottle 100g
10646271 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10209543 Supplier Thermo Scientific Acros Supplier No. 263011000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Hydroxychloroquine sulfate, CAS # 747-36-4, is a chemical compound with antiparasitic, antiviral, and anti-rheumatic activities. | CAS: 747-36-4 | C18H26ClN3O·H2SO4 | 433.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hydroxychloroquine sulfate shows antimalarial activity against malaria parasites mainly in the intra-erythrocytic phase of its life cycle. It acts by inhibiting the heme polymerase inside the food vacuole of the parasites
  • It shows an inhibitory activity in autophagy preventing the lysosome fusion with the autophagosome
  • Some studies demonstrated that hydroxychloroquine sulfate, due to its antiviral activity can restrict HIV infection in vitro

Applications

  • Hydroxychloroquine sulfate displays antiviral and anti-rheumatic activities
  • In vitro and in vivo experiments have been performed to investigate the role of hydroxychloroquine sulfate in the treatment of SARS-CoV-2 disease
  • Hydroxychloroquine sulfate can be used in immunofluorescence assays to evaluate the antiviral mechanisms in dengue virus-2 infection. In this disease, this compound can activate the innate immune response
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 747-36-4
Molecular Formula C18H26ClN3O·H2SO4
Molecular Weight (g/mol) 433.96
MDL Number MFCD00078203
InChI Key JCBIVZZPXRZKTI-UHFFFAOYSA-N
Synonym hydroxychloroquine sulfate, hydroxychloroquine sulphate, ercoquin, plaquinol, toremonil, 2-4-7-chloroquinolin-4-yl amino pentyl ethyl amino ethanol sulfate, hydroxychloroquine sulfate usp, plaquenil tn, hydroxychloroquine; sulfuric acid, dsstox_cid_27788
PubChem CID 12947
IUPAC Name 2-[4-[(7-chloroquinolin-4-yl)amino]pentyl-ethylamino]ethanol;sulfuric acid
SMILES CCN(CCCC(C)NC1=C2C=CC(=CC2=NC=C1)Cl)CCO.OS(=O)(=O)O

Specifications

Melting Point 240.0°C
Color White
Loss on Drying 2% max. (105°C, 2 hrs.)
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Glass bottle
Quantity 100 g
Merck Index 15, 4857
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol,chloroform,,ether,acetone and acetic acid
Formula Weight 433.96
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Hydroxychloroquine sulfate
Show More Show Less
compliance-icons
Product Identifier
  • Hydroxychloroquine sulfate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydroxychloroquine sulphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.