missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine O-Sulfate Sodium Salt, TRC

Product Code. 30392420
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392420 1 mg 1mg
30395708 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392420 Supplier Toronto Research Chemicals Supplier No. TRCH9169101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydroxychloroquine O-Sulfate Sodium Salt
Analyte or Component Names Hydroxychloroquine O-Sulfate Sodium Salt
Purity Grade Notes HPLC
Shipping Condition Room Temperature
Molecular Formula C18H25ClN3NaO4S
Synonym 2-[[4-[(7-Chloro-4-quinolinyl)amino]pentyl]ethylamino]ethanol 1-(Hydrogen Sulfate) Sodium Salt, 2-[[4-[(7-Chloro-4-quinolyl)amino]pentyl]ethylamino]ethanol Sulfate Sodium Salt
InChI Formula InChI=1S/C18H26ClN3O4S.Na/c1-3-22(11-12-26-27(23,24)25)10-4-5-14(2)21-17-8-9-20-18-13-15(19)6-7-16(17)18;/h6-9,13-14H,3-5,10-12H2,1-2H3,(H,20,21)(H,23,24,25);/q;+1/p-1
Solution Type Neat
SMILES ClC1=CC2=C(C(NC(C)CCCN(CC)CCOS(=O)(O[Na])=O)=CC=N2)C=C1
Industry Type Pharmaceutical
Molecular Weight (g/mol) 437.92
Quantity 1 mg
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.