missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine Acid, TRC

Product Code. 30394071
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394071 1 mg -
30403455 10 mg 10mg
30396487 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394071 Supplier Toronto Research Chemicals Supplier No. TRCH9169251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 645406-24-2
Molecular Formula C18H24ClN3O3
Molecular Weight (g/mol) 365.85
Synonym N2-(7-Chloro-4-quinolinyl)-N5-ethyl-N5-(2-hydroxyethyl)ornithine, 2-((7-Chloroquinolin-4-yl)amino)-5-(ethyl(2-hydroxyethyl)amino)pentanoic Acid
SMILES CCN(CCO)CCCC(NC1=C2C=CC(Cl)=CC2=NC=C1)C(O)=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H24ClN3O3/c1-2-22(10-11-23)9-3-4-16(18(24)25)21-15-7-8-20-17-12-13(19)5-6-14(15)17/h5-8,12,16,23H,2-4,9-11H2,1H3,(H,20,21)(H,24,25)
Formula Weight 365.1506
Chemical Name or Material Hydroxychloroquine Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.