missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxycarbamide, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Hydroxycarbamide
Specifications
Specifications
| Chemical Name or Material | Hydroxycarbamide |
| CAS | 127-07-1 |
| Analyte or Component Names | Hydroxycarbamide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C H4 N2 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-Hydroxyurea, Biosupressin, Carbamohydroxamic Acid, Carbamoyl Oxime, Cytodrox, Droxia, HU, Hidrix, Hydrea, Hydreia, Hydroxycarbamide, Hydroxycarbamine, N-(Aminocarbonyl)hydroxylamine, Hydroxylurea, Hydura, Hydurea, Litaler, Litalir, N-Carbamoylhydroxylamine, NCI C04831, NSC 32065, Oxyurea, SK 22591, SQ 1089 |
| InChI Formula | InChI=1S/CH4N2O2/c2-1(4)3-5/h5H,(H3,2,3,4) |
| SMILES | NC(=O)NO |
| IUPAC Name | hydroxyurea |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction