missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxybupropion, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Hydroxybupropion
Specifications
Specifications
| Chemical Name or Material | (±)-Hydroxybupropion |
| CAS | 92264-81-8 |
| Analyte or Component Names | (±)-Hydroxybupropion |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H18 Cl N O2 |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CC(NC(C)(C)CO)C(=O)c1cccc(Cl)c1 |
| IUPAC Name | 1-(3-chlorophenyl)-2-[(1-hydroxy-2-methylpropan-2-yl)amino]propan-1-one |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 255.74 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction