missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyalbendazole, TRC

Product Code. 30136533
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136533 50 mg 50mg
30134074 25 mg 25mg
30011872 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136533 Supplier Toronto Research Chemicals Supplier No. TRCH79720550MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107966-05-2
Molecular Formula C12H15N3O3S
Molecular Weight (g/mol) 281.33
Synonym [5-[(3-Hydroxypropyl)thio]-1H-benzimidazol-2-yl]-carbamic acid methyl ester
IUPAC Name methyl N-[5-(3-hydroxypropylsulfanyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)Nc1nc2cc(SCCCO)ccc2[nH]1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H15N3O3S/c1-18-12(17)15-11-13-9-4-3-8(7-10(9)14-11)19-6-2-5-16/h3-4,7,16H,2,5-6H2,1H3,(H2,13,14,15,17)
Formula Weight 281.0834
Chemical Name or Material Hydroxyalbendazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.