Learn More
Hydroxyacid Oxidase-1/HAO-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Products 1
Product Code Brand dimQuantity Price Quantity & Availability  
Product Code Brand dimQuantity Price Quantity & Availability  
18771454
View Documents
Novus Biologicals
NBP1-55294
£362.00
100µL
View alternative products
Save Up to 
»
Description
Description
Hydroxyacid Oxidase-1/HAO-1 Polyclonal specifically detects Hydroxyacid Oxidase-1/HAO-1 in Human samples. It is validated for Western Blot.