missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyacid Oxidase-1/HAO-1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18380896 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18380896 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18380896 Supplier Novus Biologicals Supplier No. NBP299928AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Hydroxyacid Oxidase-1/HAO-1 Polyclonal specifically detects Hydroxyacid Oxidase-1/HAO-1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hydroxyacid Oxidase-1/HAO-1
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias (S)-2-hydroxy-acid oxidase, EC 1.1.3.15, Glycolate oxidase, GOX1MGC142227, GOXMGC142225, HAOX1, hydroxyacid oxidase (glycolate oxidase) 1, hydroxyacid oxidase 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Hydroxyacid Oxidase-1/HAO-1 (NP_060015.1; Leu 2-Ile 370).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54363
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.