missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Urea-15N, TRC

Product Code. 30399458
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399458 10 mg
30404840 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399458 Supplier Toronto Research Chemicals Supplier No. TRCH99101010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214331-53-0
Molecular Formula C H4 15N N O2
Molecular Weight (g/mol) 77.05
IUPAC Name hydroxyurea
SMILES NC(=O)[15NH]O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/CH4N2O2/c2-1(4)3-5/h5H,(H3,2,3,4)/i3+1
Formula Weight 77.0243
Chemical Name or Material Hydroxyurea-15N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.