missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Tyrosol (>98%, Synthetic), TRC

Product Code. 30011423
Change view
Click to view available options
Quantity:
10 mg
50 mg
500 mg
Unit Size:
10mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011423 500 mg 500mg
30009506 50 mg 50mg
17964765 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011423 Supplier Toronto Research Chemicals Supplier No. TRCH976995500MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10597-60-1
Molecular Formula C8 H10 O3
Molecular Weight (g/mol) 154.16
Synonym Phenethyl alcohol, 3,4-dihydroxy- (6CI,7CI,8CI), 4-(2-Hydroxyethyl)-1,2-benzenediol, 1-(2-Hydroxyethyl)-3,4-dihydroxybenzene, 2-(3,4-Dihydroxyphenyl)ethanol, 2-(3,4-Dihydroxyphenyl)ethyl alcohol, 3,4-DHPEA, 3,4-Dihydroxy-β-phenethyl alcohol, 3,4-Dihydroxybenzeneethanol, 3,4-Dihydroxyphenethyl alcohol, 3,4-Dihydroxyphenylethanol, 3,4-Dihydroxyphenylethyl alcohol, 3-Hydroxytyrosol, Ba 2774, Homoprotocatechuyl alcohol, Hydroxytyrosol, β-(3,4-Dihydroxyphenyl)ethanol, β-(3,4-Dihydroxyphenyl)ethyl alcohol
IUPAC Name 4-(2-hydroxyethyl)benzene-1,2-diol
SMILES OCCc1ccc(O)c(O)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H10O3/c9-4-3-6-1-2-7(10)8(11)5-6/h1-2,5,9-11H,3-4H2
Formula Weight 154.063
Chemical Name or Material Hydroxytyrosol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.