missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Torsemide, TRC

Product Code. 30402620
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402620 5 mg
30395748 10 mg
30404075 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402620 Supplier Toronto Research Chemicals Supplier No. TRCH9699805MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99300-68-2
Molecular Formula C16H20N4O4S
Molecular Weight (g/mol) 364.42
Synonym 4-[[3-(Hydroxymethyl)phenyl]amino]-N-[[(1-methylethyl)amino]carbonyl]-3-pyridinesulfonamide, AC 3534
IUPAC Name 1-[4-[3-(hydroxymethyl)anilino]pyridin-3-yl]sulfonyl-3-propan-2-ylurea
SMILES CC(C)NC(=O)NS(=O)(=O)C1=C(C=CN=C1)NC2=CC=CC(=C2)CO

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H20N4O4S/c1-11(2)18-16(22)20-25(23,24)15-9-17-7-6-14(15)19-13-5-3-4-12(8-13)10-21/h3-9,11,21H,10H2,1-2H3,(H,17,19)(H2,18,20,22)
Formula Weight 364.1205
Chemical Name or Material Hydroxy Torsemide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.