missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Sulfentrazone, TRC

Product Code. 30398047
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398047 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30398047 Supplier Toronto Research Chemicals Supplier No. TRCH97769025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfentrazone-hydroxymethyl
CAS 134390-99-1
Purity Grade Notes HPLC
Molecular Formula C11 H10 Cl2 F2 N4 O4 S
Quantity 25 mg
Synonym Methanesulfonamide, N-[2,4-dichloro-5-[4-(difluoromethyl)-4,5-dihydro-3-(hydroxymethyl)-5-oxo-1H-1,2,4-triazol-1-yl]phenyl]-, N-[2,4-Dichloro-5-[4-(difluoromethyl)-4,5-dihydro-3-(hydroxymethyl)-5-oxo-1H-1,2,4-triazol-1-yl]phenyl]methanesulfonamide, Hydroxy Sulfentrazone, Methanesulfonamide, N-[2,4-dichloro-5-[4-(difluoromethyl)-4,5-dihydro-3-(hydroxymethyl)-5-oxo-1H-1,2,4-triazol-1-yl]phenyl]-, N-[2,4-Dichloro-5-[4-(difluoromethyl)-4,5-dihydro-3-(hydroxymethyl)-5-oxo-1H-1,2,4-triazol-1-yl]phenyl]methanesulfonamide
InChI Formula InChI=1S/C11H10Cl2F2N4O4S/c1-24(22,23)17-7-3-8(6(13)2-5(7)12)19-11(21)18(10(14)15)9(4-20)16-19/h2-3,10,17,20H,4H2,1H3
SMILES CS(=O)(=O)Nc1cc(N2N=C(CO)N(C(F)F)C2=O)c(Cl)cc1Cl
IUPAC Name N-[2,4-dichloro-5-[4-(difluoromethyl)-3-(hydroxymethyl)-5-oxo-1,2,4-triazol-1-yl]phenyl]methanesulfonamide
Molecular Weight (g/mol) 403.189
Formula Weight 401.977
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.