missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Saxagliptin, TRC

Product Code. 30388874
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388874 1 mg -
30405513 5 mg 5mg
30397345 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388874 Supplier Toronto Research Chemicals Supplier No. TRCH9534651MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 841302-24-7
Molecular Formula C18 H25 N3 O3
Molecular Weight (g/mol) 331.41
Synonym 2-Azabicyclo[3.1.0]hexane-3-carbonitrile, 2-[(2S)-2-amino-2-(3,5-dihydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-, (1S,3S,5S)-, 2-Azabicyclo[3.1.0]hexane-3-carbonitrile, 2-[(2S)-amino(3,5-dihydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-, (1S,3S,5S)- (9CI), (1S,3S,5S)-2-[(2S)-2-Amino-2-(3,5-dihydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile, 5-Hydroxy saxagliptin, BMS 510849, M2 saxagliptin hydroxylated metabolite
IUPAC Name (1S,3S,5S)-2-[(2S)-2-amino-2-(3,5-dihydroxy-1-adamantyl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile
SMILES N[C@H](C(=O)N1[C@@H](C[C@@H]2C[C@H]12)C#N)C34CC5CC(O)(CC(O)(C5)C3)C4

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C18H25N3O3/c19-6-12-1-11-2-13(11)21(12)15(22)14(20)16-3-10-4-17(23,7-16)9-18(24,5-10)8-16/h10-14,23-24H,1-5,7-9,20H2/t10?,11-,12+,13+,14-,16?,17?,18?/m1/s1
Formula Weight 331.1896
Chemical Name or Material hydroxy Saxagliptin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.