missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Pioglitazone (M-VII), TRC

Product Code. 30389080
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389080 5 mg -
30405529 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30389080 Supplier Toronto Research Chemicals Supplier No. TRCH9520605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625853-72-7
Molecular Formula C19H20N2O4S
Molecular Weight (g/mol) 372.44
Synonym 5-[[4-[2-[5-(2-Hydroxyethyl)-2-pyridinyl]ethoxy]phenyl]methyl]-2,4-thiazolidinedione
IUPAC Name 5-[[4-[2-[5-(2-hydroxyethyl)pyridin-2-yl]ethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
SMILES OCCc1ccc(CCOc2ccc(CC3SC(=O)NC3=O)cc2)nc1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C19H20N2O4S/c22-9-7-14-1-4-15(20-12-14)8-10-25-16-5-2-13(3-6-16)11-17-18(23)21-19(24)26-17/h1-6,12,17,22H,7-11H2,(H,21,23,24)
Formula Weight 372.1144
Chemical Name or Material Hydroxy Pioglitazone (M-VII)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.