missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Pioglitazone (M-II) (Mixture of Diastereomers), TRC

Product Code. 30391265
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30391265 100 mg 100mg
30398082 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30391265 Supplier Toronto Research Chemicals Supplier No. TRCH952080100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101931-00-4
Molecular Formula C19 H20 N2 O4 S
Molecular Weight (g/mol) 372.44
Synonym 2,4-Thiazolidinedione, 5-[[4-[2-(5-ethyl-2-pyridinyl)-2-hydroxyethoxy]phenyl]methyl]-, 5-[[4-[2-(5-Ethyl-2-pyridyl)-2-hydroxy-ethoxy]phenyl]methyl]thiazolidine-2,4-dione
IUPAC Name 5-[[4-[2-(5-ethylpyridin-2-yl)-2-hydroxyethoxy]phenyl]methyl]-1,3-thiazolidine-2,4-dione
SMILES CCc1ccc(nc1)C(O)COc2ccc(CC3SC(=O)NC3=O)cc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C19H20N2O4S/c1-2-12-5-8-15(20-10-12)16(22)11-25-14-6-3-13(4-7-14)9-17-18(23)21-19(24)26-17/h3-8,10,16-17,22H,2,9,11H2,1H3,(H,21,23,24)
Formula Weight 372.1144
Chemical Name or Material Hydroxy Pioglitazone (M-II) (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.