missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Naphthol Blue Disodium Salt (Technical Grade), TRC

Product Code. 30392925
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392925 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392925 Supplier Toronto Research Chemicals Supplier No. TRCH95109050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 165660-27-5
Molecular Formula C20 H12 N2 O11 S3 . 2Na
Quantity 50 g
Synonym 3-Hydroxy-4-[(2-hydroxy-4-sulfo-1-naphthalenyl)azo]-2,7-naphthalenedisulfonic Acid Disodium Salt
InChI Formula InChI=1S/C20H14N2O11S3.2Na/c23-15-9-16(35(28,29)30)13-3-1-2-4-14(13)18(15)21-22-19-12-6-5-11(34(25,26)27)7-10(12)8-17(20(19)24)36(31,32)33;;/h1-9,23-24H,(H,25,26,27)(H,28,29,30)(H,31,32,33);;/q;2*+1/p-2
SMILES [Na+].[Na+].Oc1cc(c2ccccc2c1N=Nc3c(O)c(cc4cc(ccc34)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)O
IUPAC Name disodium;3-hydroxy-4-[(2-hydroxy-4-sulfo-1-naphthyl)azo]naphthalene-2,7-disulfonate
Molecular Weight (g/mol) 598.49062
Formula Weight 597.9399
Chemical Name or Material Hydroxy Naphthol Blue Disodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.