missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy naphthol blue disodium salt, For ACS analysis, MP Biomedicals™

Product Code. 11422852 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422852 10 g 10g
11432852 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11422852 Supplier MP Biomedicals Supplier No. 0215252910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 165660-27-5
Molecular Formula C20H14N2Na2O11S3
Molecular Weight (g/mol) 600.495
InChI Key LXJHFXHZWYBBKJ-LUUCHEBKSA-N
Synonym `-(2-naphtholazo-3, 6-disulfonic acid)-2-naphthol-4-sulfonic acid trisodium salt, calcium indicator on sodium chloride
PubChem CID 131849366
IUPAC Name 3-hydroxy-4-[(2Z)-2-(2-oxo-4-sulfonaphthalen-1-ylidene)hydrazinyl]naphthalene-2,7-disulfonic acid;sodium
SMILES C1=CC=C2C(=C1)C(=CC(=O)C2=NNC3=C4C=CC(=CC4=CC(=C3O)S(=O)(=O)O)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Specifications

Formula Weight 598.5
Color Black to Blue
Quantity 10 g
Physical Form Powder
Chemical Name or Material Hydroxy naphthol blue disodium salt, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.